DWK Life Sciences Kimble™ Vacuum Desiccator with Collar, 250 x 330
Ideal for removing moisture from solids, storage of hygroscopic chemicals, and applications requiring vacuum desiccation.
Supplier:  DWK Life Sciences DWK31200250
Catalog No. 12-141-435
Description
Description
  • Greased rotating collar valve is used to facilitate vacuum desiccation
  • Includes solid glass lid with collar valve, metal plate, and a recessed base for desiccant containment
  • Manufactured from soda-lime glass conforming to USP Type III and ASTM E438, Type II requirements
  • Can be used for cooling oven-dried samples while maintaining sample dryness
Specifications
Specifications
Vacuum Desiccator with Collar
SDS
Documents
Documents
Accessories