GelDoc-It™ DNA and Protein Gel Imaging System
Manufacturer:  UVP Inc 97057701
 View more versions of this product
Catalog No. UVP97057701
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
Windows™ 7, XP, Vista
0 to 65, 535
1280 x 1024
1.3MP, extendable to 5MP
14.5 in.
DNA and protein gel imaging, blue light gels, TLC gels, colony plates, visible light gels, autoradiograph: fluorescence, colorimetric
85.1cm
VisionWorksLS image acquisition and analysis, single-user license
44.4cm
Ethidium Bromide Broadband
8 to 48mm for 1.2 manual zoom
GelDoc-Ite Imaging System
UV: 254/302/365nm
36.8cm
33.5 in.
FluorCam 220 camera/lens kit, darkroom with gel viewing window, epi white light, Ethidium Bromide filter, choice of transilluminator and VisionWorksLS capture and analysis software
17.5 in.
Documents