Upchurch Scientific™ MicroTight™ One-piece Fittings DFS Item
Manufacturer:  Upchurch Scientific™ UPF125X
 View more versions of this product
Catalog No. 29-008-323
Specifications
Specifications
0.53 in.
0.31 in.
Each
One Piece Fitting
4000psi
13.7mm
7.87mm
PEEK
Natural
Documents