missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ UltraComp eBeads™ Compensation Beads

Designed for use in compensation with all fluorochromes excited by ultraviolet, violet, blue, green, yellow-green, and red lasers. UltraComp eBeads™ react with antibodies of mouse, rat, and hamster origin, and are immunoglobulin light chain-independent. 1 drop (50μL)/test.

$178.00 - $491.00

Specifications

Product Line eBeads™
Product Type Compensation Beads
For Use With (Equipment) Flow Cytometer
View More Specs

Alert:

Contains Sodium azide


Products 2
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
501129040
View Documents Promotion Details
Invitrogen™
01222242
Each for $491.00
 
501129038
View Documents Promotion Details
Invitrogen™
01222241
Each for $178.00
 
Description

Description

  • Each drop of beads contains both a positive population that will capture any mouse, rat or hamster antibody and a negative population that will not react with antibody
  • This bimodal distribution can be used for single-color compensation controls in multicolor flow cytometry experiments
  • Compatible with all fluorochromes excited by an ultraviolet (355nm) or violet (405nm) laser
  • Should be used with standard staining buffers which contain PBS or HBSS, protein such as BSA or FBS, and sodium azide. No other additives should be used.
Specifications

Specifications

eBeads™
Compensation Beads
Flow Cytometer
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.