missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ Tyvek™ IsoClean™ Series 253 Bound Seam Coveralls, Clean-Processed
Clean-processed coveralls provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash. Bound seams reduce the potential for particle penetration.
$448.61 - $587.91
Specifications
Product Type | Cleanroom Coveralls |
---|---|
Material | Flashspun Polyethylene |
Color | White |
Disposable | Single-use |
Ankle Style | Covered Elastic |
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Catalog Number | Mfr. No. | Size | Height (English) | Height (Metric) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Height (English) | Height (Metric) | Price | Quantity | |||
17-990-511
|
DuPont™
IC253BWHSM00250C |
Small | 5 to 5.7 ft. | 152.4 to 173.7 cm |
Case of 25 Each for $551.34
|
|
|||
17-988-034
|
DuPont™
IC253BWH2X00250C |
2X-Large | 6.1 to 6.5 ft. | 185.92 to 198.12 cm |
Case of 25 Each for $475.51
|
|
|||
17-988-032
|
DuPont™
IC253BWHLG00250C |
Large | 5.8 to 6 ft. | 176.78 to 182.88 cm |
Case of 25 Each for $448.61
|
|
|||
17-988-035
|
DuPont™
IC253BWH3X00250C |
3X-Large | 6.2 to 6.6 ft. | 188.97 to 201.16 cm |
Case of 25 Each for $504.04
|
|
|||
17-988-031
|
DuPont™
IC253BWHMD00250C |
Medium | 5.5 to 5.10 ft. | 167.64 to 155.44 cm |
Case of 25 Each for $448.61
|
|
|||
19-188-130
|
DuPont™
IC253BWH5X00250C |
5X-Large | 6.4 to 7.2 ft. | 195.07 to 219.45 cm |
Case of 25 Each for $587.91
|
|
|||
17-988-036
|
DuPont™
IC253BWH4X00250C |
4X-Large | 6.3 to 7 ft. | 192.02 to 213.3 cm |
Case of 25 Each for $544.33
|
|
|||
17-988-033
|
DuPont™
IC253BWHXL00250C |
X-Large | 5.11 to 6.3 ft. | 155.75 to 192.04 cm |
Case of 25 Each for $448.61
|
|
|||
Description
- Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio
- Bound seams are covered with garment fabric to reinforce the seam
- Elastic openings for tighter fit at wrist and ankle
- Bound neck for lower particle shedding
- Front zipper closure for easy donning and doffing
- Dolman sleeve design for greater range of motion and comfort
Specifications
Cleanroom Coveralls | |
White | |
Covered Elastic | |
Collared | |
Bound | |
Bound | |
5 mil | |
Clean-processed |
Flashspun Polyethylene | |
Single-use | |
Covered Elastic | |
Zipper | |
Non-sterile | |
Dolman | |
Barrier to dry particles, microorganisms, and non-hazardous liquids in controlled environments | |
Class 1000 (ISO 6) |