missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ Tyvek™ IsoClean™ Hood with Attached Mask
Delivers an ideal balance of protection, durability, and comfort.
Supplier: DuPont™ IC669BWH0001000S
Description
- Hood with attached 7 in. blue mask
- Made using a patented flash spinning process, provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash
- Pleated outer mask is made of polyethylene
- Seams and head opening are bound; ties and loops provide a secure fit
- Sterile garments are gamma sterilized to SAL of 10-6
- Packaged in an ISO Class 5 cleanroom
- Individually packaged in an opaque bag and placed in double transparent poly liners
Specifications
Hood With Attached Mask | |
Tyvek | |
Ties with Loops | |
Bound | |
ISO Class 5 cleanroom | |
18.375 in. | |
Polyethylene 7 in. Blue Mask |
Universal | |
White | |
IC669B | |
Sterile | |
Individually packaged in an opaque bag and placed in double transparent poly liners | |
29.625 in. | |
Class 1-10 (ISO 3-4) |