Learn More
Tylosin tartrate, 94%, Thermo Scientific Chemicals
Macrolide antibiotic
$98.58 - $646.07
Chemical Identifiers
CAS | 1405-54-5 |
---|---|
Molecular Formula | C50H83NO23 |
Molecular Weight (g/mol) | 1066.198 |
MDL Number | MFCD00070094 |
InChI Key | ICVKYYINQHWDLM-HYHSIFHGSA-N |
Synonym | Pharmasin; [R-(R'R')]-2,3-Dihydroxybutanedionate Tylosin salt |
PubChem CID | 131673832 |
IUPAC Name | (2R,3R)-2,3-dihydroxybutanedioic acid;2-[(4R,5S,6S,7R,9R,15R,16R)-6-[(2R,3R,4R,5S,6R)-5-[(2S,4R,5S,6S)-4,5-dihydroxy-4,6-dimethyloxan-2-yl]oxy-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-16-ethyl-4-hydroxy-15-[[(2R,3R,4R,5R,6R)-5-hydroxy-3,4-dimetho |
SMILES | CCC1C(C=C(C=CC(=O)C(CC(C(C(C(CC(=O)O1)O)C)OC2C(C(C(C(O2)C)OC3CC(C(C(O3)C)O)(C)O)N(C)C)O)CC=O)C)C)COC4C(C(C(C(O4)C)O)OC)OC.C(C(C(=O)O)O)(C(=O)O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6263303
|
Thermo Scientific Chemicals
J6263303 |
1 g |
Each for $98.58
|
|
|||||
AAJ6263306
|
Thermo Scientific Chemicals
J6263306 |
5 g |
Each for $333.97
|
|
|||||
AAJ6263309
|
Thermo Scientific Chemicals
J6263309 |
10 g |
Each for $646.07
|
|
|||||
Description
It is used as an antibacterial agent. This compound has been shown to be effective in vitro against Staphylococcus intermedius and sensitive mycoplasma organisms. Although a lot of antibiotics tend to suppress the immune system, macrolide antibiotics have been reported to enhance the immune system when used in chicken models. Studies suggest that chronic exposure to this compound may have a negative effect on the respiratory system. For turkeys - for treatment of infectious synovitis and sinusitis. It is effective also for treatment of ileitis, dysentery and necrotic enteritis in swine. It is successfully applied as a preventive measure in stress conditions as vaccinations, transportation and others.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as an antibacterial agent. This compound has been shown to be effective in vitro against Staphylococcus intermedius and sensitive mycoplasma organisms. Although a lot of antibiotics tend to suppress the immune system, macrolide antibiotics have been reported to enhance the immune system when used in chicken models. Studies suggest that chronic exposure to this compound may have a negative effect on the respiratory system. For turkeys - for treatment of infectious synovitis and sinusitis. It is effective also for treatment of ileitis, dysentery and necrotic enteritis in swine. It is successfully applied as a preventive measure in stress conditions as vaccinations, transportation and others.
Solubility
Soluble in water.
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Chemical Identifiers
1405-54-5 | |
1066.198 | |
ICVKYYINQHWDLM-HYHSIFHGSA-N | |
131673832 | |
CCC1C(C=C(C=CC(=O)C(CC(C(C(C(CC(=O)O1)O)C)OC2C(C(C(C(O2)C)OC3CC(C(C(O3)C)O)(C)O)N(C)C)O)CC=O)C)C)COC4C(C(C(C(O4)C)O)OC)OC.C(C(C(=O)O)O)(C(=O)O)O |
C50H83NO23 | |
MFCD00070094 | |
Pharmasin; [R-(R'R')]-2,3-Dihydroxybutanedionate Tylosin salt | |
(2R,3R)-2,3-dihydroxybutanedioic acid;2-[(4R,5S,6S,7R,9R,15R,16R)-6-[(2R,3R,4R,5S,6R)-5-[(2S,4R,5S,6S)-4,5-dihydroxy-4,6-dimethyloxan-2-yl]oxy-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-16-ethyl-4-hydroxy-15-[[(2R,3R,4R,5R,6R)-5-hydroxy-3,4-dimetho |
Specifications
Tylosin Tartrate | |
1405-54-5 | |
MFCD00070094 | |
Soluble in water. | |
CCC1C(C=C(C=CC(=O)C(CC(C(C(C(CC(=O)O1)O)C)OC2C(C(C(C(O2)C)OC3CC(C(C(O3)C)O)(C)O)N(C)C)O)CC=O)C)C)COC4C(C(C(C(O4)C)O)OC)OC.C(C(C(=O)O)O)(C(=O)O)O | |
1066.198 | |
1066.2 | |
Cream to White | |
Tylosin tartrate |
Powder | |
C50H83NO23 | |
Pharmasin; [R-(R'R')]-2,3-Dihydroxybutanedionate Tylosin salt | |
ICVKYYINQHWDLM-HYHSIFHGSA-N | |
(2R,3R)-2,3-dihydroxybutanedioic acid;2-[(4R,5S,6S,7R,9R,15R,16R)-6-[(2R,3R,4R,5S,6R)-5-[(2S,4R,5S,6S)-4,5-dihydroxy-4,6-dimethyloxan-2-yl]oxy-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-16-ethyl-4-hydroxy-15-[[(2R,3R,4R,5R,6R)-5-hydroxy-3,4-dimetho | |
131673832 | |
95% | |
1 g |
Safety and Handling
GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
H317-H334
EINECSNumber : 215-781-5
RTECSNumber : YP0875000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only