missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tween 20™, Ultrapure, Thermo Scientific Chemicals
Supplier: Thermo Fisher Scientific J20605AP
Specifications
Tween 20™ | |
Liquid | |
1.105 | |
MFCD00165986 | |
Polyoxyethylene sorbitan monolaurate | |
HMFKFHLTUCJZJO-UHFFFAOYNA-N | |
2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate | |
443314 | |
Plastic bottle |
9005-64-5 | |
>100°C | |
(C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 | |
500 mL | |
>110°C | |
CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO | |
522.68 | |
Ultra Pure | |
1.105 |
Chemical Identifiers
9005-64-5 | |
522.68 | |
HMFKFHLTUCJZJO-UHFFFAOYNA-N | |
443314 |
(C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 | |
MFCD00165986 | |
Polyoxyethylene sorbitan monolaurate | |
CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
Safety and Handling
EINECSNumber : 500-018-3
RTECSNumber : TR7400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only