Eppendorf™ Tube Adapters for Eppendorf™ Model 5702 Round Buckets
Fit in round bucket 05-400-320 to accept tubes from 1.1 to 85mL
Supplier:  Eppendorf™ 022639285
 View more versions of this product
Catalog No. 05-400-326
Description
Description
Adapters are sold in packs of two. Be sure to order proper number to fully equip your rotor.
Specifications
Specifications
A-4-38 swing bucket rotor (round bucket) for 5702/5702R/5702RH centrifuge
1.1 to 1.4 mL
5