Eppendorf™ Tube Adapters for Eppendorf™ Eight-Place Swinging Bucket Rotor
For Eppendorf 5702 centrifuges
$80.73 - $276.10
Specifications
Type | Tube Adapter |
---|---|
For Use With (Equipment) | Eppendorf™ 5702 centrifuge |
Catalog Number | Mfr. No. | Tube Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Tube Type | Price | Quantity | |||||
05-400-302
|
Eppendorf™
022639510 |
Round Bottom |
Pack of 8 Each for $80.73
|
|
|||||
05-400-303
|
Eppendorf™
022639528DUP |
Conical |
Pack of 8 Each for $276.10
|
|
|||||
Description
Fit in bucket supplied with 05-400-301 swinging bucket rotor to accept 15mL glass or conical tubes.
Specifications
Tube Adapter | |
Eppendorf™ 5702 centrifuge |