missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
β-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate, 97+%, Thermo Scientific Chemicals
$174.06 - $1439.84
Chemical Identifiers
CAS | 698999-85-8 |
---|---|
Molecular Formula | C21H27N7NaO17P3 |
Molecular Weight (g/mol) | 765.39 |
MDL Number | MFCD00036973 |
InChI Key | JNUMDLCHLVUHFS-QYZPTAICSA-M |
Synonym | nadp sodium salt, beta-nicotinamide adenine dinucleotide phosphate sodium |
PubChem CID | 131845262 |
SMILES | [Na+].NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP(O)(O)=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC227571000
|
Thermo Scientific Chemicals
227571000 |
100 mg | Glass bottle |
Each for $174.06
|
|
||||
AC227575000
|
Thermo Scientific Chemicals
227575000 |
500 mg | Glass bottle |
Each for $371.18
|
|
||||
AC227570025
|
Thermo Scientific Chemicals
227570025 |
2.5 g | Glass bottle |
Each for $1,439.84
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
698999-85-8 | |
765.39 | |
JNUMDLCHLVUHFS-QYZPTAICSA-M | |
131845262 |
C21H27N7NaO17P3 | |
MFCD00036973 | |
nadp sodium salt, beta-nicotinamide adenine dinucleotide phosphate sodium | |
[Na+].NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP(O)(O)=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O |
Specifications
175.0°C to 178.0°C | |
698999-85-8 | |
100 mg | |
C21H27N7NaO17P3 | |
14,6348 | |
Solubility in water: soluble. Other solubilities: soluble in methanol,less soluble in ethanol,practically insoluble in ether and ethyl acetate | |
[Na+].NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP(O)(O)=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O | |
765.39 | |
765.39 | |
Authentic | |
β-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate |
White to Yellow | |
Powder | |
determination,based on dry weight) | |
MFCD00036973 | |
nadp sodium salt, beta-nicotinamide adenine dinucleotide phosphate sodium | |
JNUMDLCHLVUHFS-QYZPTAICSA-M | |
[[(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate;sodium | |
131845262 | |
≥97% | |
Glass bottle |
Safety and Handling
EINECSNumber : 214-664-6
RUO – Research Use Only