missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ TraceableLIVE™ Wi-Fi Datalogging Refrigerator/Freezer Thermometers with Remote Notification

The simple, efficient, and reliable way to stay connected to critical environments through a smart phone, tablet, or personal computer.

$658.94 - $729.56

Specifications

Temperature Range (Metric) -50 to +60°C
Accuracy (Temperature) ±0.25°C
Temperature Resolution 0.01°
View More Specs

Includes: Two probes, quick start guide, batteries, magnets, Velcro, wall mount, and Traceable™ certificate

Products 4
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
15-079-615 View Documents Fisherbrand™
6500**
2 Bottles (patented) Each for $682.47
15-079-616 View Documents Fisherbrand™
6501**
1 Bottle & 1 Bullet™ probe Each for $673.06
15-079-617 View Documents Fisherbrand™
6502**
2 Bullet™ probes Each for $658.94
15-079-618 View Documents Fisherbrand™
6503
2 Stainless-steel probes Each for $729.56
Description

Description

The simple, efficient, and reliable way to stay connected to critical environments through a smart phone, tablet, or personal computer.

Using the TraceableLIVE™ app or visiting Traceablelive.com on any browser allows the user to view current temperatures, control alarm parameters, view datalogging history, view and generate reports, and give other team members access to any TraceableLIVE™ equipment at any time.

Features:
  • Monitor temperature and more either locally or remotely — connect to existing Wi-Fi
  • Alerts: temperature alarm, connectivity interruption, low battery
  • Cloud-based data interface means no local software required; web, Android, Apple IOS data visibility
  • Continuous data transmission and monitoring using SSL encrypted SECURE connection
  • Receive MOBILE PUSH / E-MAIL / TEXT notifications
  • Unlimited scalability — use TraceableLIVE™ in individual labs or an entire network of facilities
  • Multiple point temperature calibration — traceable to NIST
Note:  Requires TraceableLIVE™ subscription, starting at $8.99/month per device.

Certifications

Includes certificate assuring accuracy from an ISO/IEC 17025:2005 (1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI).

Specifications

Specifications

-50 to +60°C
±0.25°C
0.01°