missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Total Organic Carbon (TOC), standard solution, Specpure™ 1000μg/mL, Thermo Scientific Chemicals
$219.96 - $433.01
Specifications
Chemical Name or Material | Total Organic Carbon |
---|---|
Name Note | 1000μg/mL |
Color | Colorless |
Molecular Formula | KHC8H4O4 in H2O |
MDL Number | MFCD00133992 |
Description
Total Organic Carbon (TOC), is used as a standard solution during the measurement of chemical analysis such as detecting contaminants in drinking water and cooling water. It is also used for analysis of water to be used for semiconductor manufacturing and pharmaceutical use.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Total Organic Carbon | |
Colorless | |
MFCD00133992 | |
Miscible with water |
1000μg/mL | |
KHC8H4O4 in H2O | |
Characteristic | |
Liquid |
RUO – Research Use Only