missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Toluene, Optima™, Fisher Chemical™
High purity and versatile solvent suitable for HPLC, spectrophotometry, and pesticide residue analysis
$221.67 - $572.42
Chemical Identifiers
CAS | 108-88-3 |
---|---|
Molecular Formula | C7H8 |
Molecular Weight (g/mol) | 92.14 |
MDL Number | MFCD00008512 |
InChI Key | YXFVVABEGXRONW-UHFFFAOYSA-N |
Synonym | methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene |
PubChem CID | 1140 |
ChEBI | CHEBI:17578 |
IUPAC Name | toluene |
SMILES | CC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
T290-1
|
Fisher Chemical
T2901 |
1 L | Glass Bottle |
Each for $221.67
Case of 6 Each for $1,330.02
|
|
||||
T291-4
|
Fisher Chemical
FLT2914 |
4 L | Amber Glass |
Each for $572.42
Case of 4 Each for $2,289.68
|
|
||||
Description
- Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
- Tested by GC-ECD to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis
- Also meets ACS specifications
- Intended for research use only
Chemical Identifiers
108-88-3 | |
92.14 | |
YXFVVABEGXRONW-UHFFFAOYSA-N | |
1140 | |
toluene |
C7H8 | |
MFCD00008512 | |
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene | |
CHEBI:17578 | |
CC1=CC=CC=C1 |
Specifications
-95°C | |
1 AU max. at 286nm | |
0.866g/cm³ | |
1 L | |
Glass Bottle | |
1.4930 to 1.4950 (at 25°C) | |
99.8% min by GC | |
MFCD00008512 | |
0.6 mPa/s at 20°C | |
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene | |
CC1=CC=CC=C1 | |
92.14 | |
CHEBI:17578 | |
≥99.8% | |
Toluene, Also meets ACS specifications |
108-88-3 | |
Colorless | |
111°C | |
Pass Test | |
Liquid | |
10ppm max. | |
C7H8 | |
29mbar at 20°C | |
0.02% max. | |
YXFVVABEGXRONW-UHFFFAOYSA-N | |
toluene | |
1140 | |
92.14 | |
Mobile phase for HPLC applications |
Safety and Handling
DANGER!
Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0
Signal Word: DANGER!
DOTInformation : DOT Class 3, : Flammable Liquid