missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Toluene, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.8%, Honeywell
$488.23 - $3543.48
Chemical Identifiers
CAS | 108-88-3 |
---|---|
Molecular Formula | C7H8 |
Molecular Weight (g/mol) | 92.14 |
MDL Number | MFCD00008512 |
InChI Key | YXFVVABEGXRONW-UHFFFAOYSA-N |
Synonym | methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene |
PubChem CID | 1140 |
ChEBI | CHEBI:17578 |
IUPAC Name | toluene |
SMILES | CC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
6001724
|
Honeywell Chemicals
LP3474 |
4 x 4 L | Glass Bottle |
Case of 4 Each for $577.63
|
|
||||
6001722
|
Honeywell Inc
LP34719 |
19 L | Metal Can |
Each for $488.23
|
|
||||
6001723
|
Honeywell Chemicals
LP347204 |
204 L | Metal Drum |
Each for $3,543.48
|
|
||||
Chemical Identifiers
108-88-3 | |
92.14 | |
YXFVVABEGXRONW-UHFFFAOYSA-N | |
1140 | |
toluene |
C7H8 | |
MFCD00008512 | |
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene | |
CHEBI:17578 | |
CC1=CC=CC=C1 |
Specifications
108-88-3 | |
0.867 g/cm3 | |
4 x 4 L | |
Glass Bottle | |
MFCD00008512 | |
0.03% max. | |
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene | |
YXFVVABEGXRONW-UHFFFAOYSA-N | |
toluene | |
1140 | |
92.14 | |
ACS Reagent | |
by GC Analysis |
100.0000% | |
110.6°C | |
4°C | |
C7H8 | |
UN1294 | |
635760 | |
5.2% in Water at 20°C | |
CC1=CC=CC=C1 | |
92.14 | |
CHEBI:17578 | |
>99.5% | |
Toluene, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.8% |
Safety and Handling
ShelfLife : 547 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Toluene