missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals Tioconazole, 98+%
Potent inhibitor of cytochrome-P450
$274.32 - $439.70
Chemical Identifiers
CAS | 65899-73-2 |
---|---|
Molecular Formula | C16H13Cl3N2OS |
Synonym | Thioconazole |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6045903
|
Thermo Scientific Chemicals
J6045903 |
1 g |
Each for $274.32
|
|
|||||
AAJ6045906
|
Thermo Scientific Chemicals
J6045906 |
5 g |
Each for $439.70
|
|
|||||
Description
Potent inhibitor of cytochrome-P450
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
65899-73-2 | |
C16H13Cl3N2OS | |
Thioconazole |
Specifications
1 g | |
Potent inhibitor of cytochrome-P450 | |
C16H13Cl3N2OS | |
Thioconazole | |
Powder |
White to Off-white | |
65899-73-2 | |
387.71g/mol | |
Signal Transduction Reagent | |
≥98% |
Safety and Handling
EINECSNumber : 265-973-8
Recommended Storage : Ambient temperatures
RUO – Research Use Only