missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tin(IV) 2-ethylhexanoate diisopropoxide, 98% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03916903

 View more versions of this product

Catalog No. AA3916903



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tin(IV) 2-ethylhexanoate diisopropoxide
(metals basis)
1 g
tin iv 2-ethylhexanoate diisopropoxide, tin iv 2-ethylhexanoatediisopropoxide metalsbasis
98%
288374-32-3
MFCD00210604
Moisture sensitive
523.08
Liquid

Chemical Identifiers

288374-32-3
MFCD00210604
tin iv 2-ethylhexanoate diisopropoxide, tin iv 2-ethylhexanoatediisopropoxide metalsbasis
Safety and Handling

Safety and Handling

GHS H Statement
H361-H315-H319
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.

P201-P202-P264b-P280i-P281-P302+P352-P305+P351+P338-P308+P313-P332+P313-P362-P501c

H315-H319-H361d

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only