Learn More
Tin(II) oxalate, Thermo Scientific Chemicals
$58.27 - $204.53
Chemical Identifiers
CAS | 814-94-8 |
---|---|
Molecular Formula | C2O4Sn |
Molecular Weight (g/mol) | 206.728 |
MDL Number | MFCD00040678 |
InChI Key | OQBLGYCUQGDOOR-UHFFFAOYSA-L |
Synonym | stannous oxalate, tin ii oxalate, tin oxalate, tin 2+ oxalate, ethanedioic acid, tin 2+ salt 1:1, stavelan cinaty czech, unii-sar72fe8eh, sar72fe8eh, stavelan cinaty |
PubChem CID | 13149 |
IUPAC Name | oxalate;tin(2+) |
SMILES | C(=O)(C(=O)[O-])[O-].[Sn+2] |
Description
Tin(II) oxalate is used for dyeing and printing textiles. It acts as a catalyst for the esterification reactions between long chain fatty acids and alcohols and in polymerization reactions. It is also used in stannous oral care compositions. Further, it is used in proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
814-94-8 | |
206.728 | |
OQBLGYCUQGDOOR-UHFFFAOYSA-L | |
13149 | |
C(=O)(C(=O)[O-])[O-].[Sn+2] |
C2O4Sn | |
MFCD00040678 | |
stannous oxalate, tin ii oxalate, tin oxalate, tin 2+ oxalate, ethanedioic acid, tin 2+ salt 1:1, stavelan cinaty czech, unii-sar72fe8eh, sar72fe8eh, stavelan cinaty | |
oxalate;tin(2+) |
Specifications
814-94-8 | |
White | |
C2O4Sn | |
MFCD00040678 | |
UN3288 | |
14,8786 | |
Soluble in dilute HCl; Insoluble in water;Soluble in acids; Insoluble in water and acetone. | |
C(=O)(C(=O)[O-])[O-].[Sn+2] | |
206.728 | |
206.72 | |
Tin(II) oxalate |
280°C (decomposition) | |
3.56 g/mL | |
SnC2O4 | |
100 g | |
3708588 | |
stannous oxalate, tin ii oxalate, tin oxalate, tin 2+ oxalate, ethanedioic acid, tin 2+ salt 1:1, stavelan cinaty czech, unii-sar72fe8eh, sar72fe8eh, stavelan cinaty | |
OQBLGYCUQGDOOR-UHFFFAOYSA-L | |
oxalate;tin(2+) | |
13149 | |
Powder |
Safety and Handling
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber : 212-414-0
RTECSNumber : XQ3950000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only