Thermo Scientific™ ThermoChill II Recirculating Chiller
Reliable liquid temperature control, with cooling capacity up to 1000W/3415 BTU.
Manufacturer:  Thermo Scientific™ 197112010000
Catalog No. 13-875-102
Description
Description
  • Minimized plumbing connections
  • Integrated full-flow fluid strainer
  • Compact, simple design and operation
Specifications
Specifications
1000w
115V
23.8 in.
50Hz
120 lb.
+/- 0.1°C
35.6cm
23 in.
R134A
2.5 gal.
115V 50Hz
11.3L
2.5 gpm @ 60 psid
60.5cm
PD2 (Positive Displacement Pump)
54.4kg
14 in.
10° to 35°C
58.4cm
+5 to +30°C
CE markedUL listed
Documents