Thermo Scientific™ TRACE™ 1300 GC Startup Package
A versatile and budget-friendly investment for the basic routine laboratory
Manufacturer:  Thermo Scientific 1205100110
Catalog No. 03-950-151
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
The TRACE 1300 Series Gas Chromatograph is the latest technology breakthrough conceived to substantially elevate performance in QA/QC and routine laboratories. Developed around key innovations driven by customer needs, including user-exchangeable instant connect injectors and detectors, enhanced robustness of components, scaling-down of injector and detector solutions, immediate local user interface, and optimization of all electronic elements, the TRACE 1300 Series GC results in an extremely fast, easy to use, compact GC, delivering an incredibly high lab productivity at a reduced cost of ownership.
Ultimate Productivity
  • Easy adoption of standard GC methods
  • Increased robustness of injector technology
  • Shorter sample cycle time
  • Unmatched detector sensitivity in trace analysis
  • Scalable, simply intelligent chromatography data system
Reduced cost of ownership
  • Tailorable GC configurations
  • Smart maintenance
  • Consumable adaptability
  • Seamless automation
  • Energy-saving
  • Minimal sample and solvent consumption
Features a simplified user interface with single button start/stop and maintenance operations while offering excellent features and capabilities.
Startup Package also includes:
Thermo Scientific Dionex™ Chromeleon™ 7.1
  • Enjoy Operational Simplicity - See how Chromeleon 7.1's thoughtfully designed user interface and visual cues guide you naturally through your chromatography
  • Streamline Your Workflow - Simplify your analysis to just a few mouse clicks, without sacrificing flexibility, using the innovative eWorkflows feature
  • Speed Up Data Analysis - Discover how much time you can save with Chromeleon 7.1's Cobra Peak Detection algorithm, SmartPeaks™ Integration Assistant, and dynamic interactive data processing
  • Multiple Instrument Control - Use Chromeleon 7.1 to unify control of all your instruments, regardless of type (LC/IC/GC) or vendor
  • Ensure Compliance - Satisfy regulatory requirements without sacrificing efficiency using Chromeleon 7.1's integrated security system, audit trails, and version management tools
  • Deliver Results - Get the most out of your data quickly and easily, with powerful data mining tools and a built-in spreadsheet that updates dynamically
Installation/Start-up Kit
Documents