Thermo Scientific™ Sorvall™ Adapters for Stratos™ Four-Place Swinging Bucket Rotor
For use with Thermo Scientific Sorvall Stratos Centrifuges in Four-Place Swinging Bucket Rotor 75-003-047 with Round Bucket 75-008-172
$228.04 - $244.40
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
75-008-183
|
Thermo Scientific™
75008183 |
Green/Yellow |
Each for $244.40
|
|
|||||
75-008-182
|
Thermo Scientific™
75008182 |
Green |
Each for $228.04
|
|
|||||
75-008-187
|
Thermo Scientific™
75008187 |
Brown |
Each for $231.67
|
|
|||||
75-008-185
|
Thermo Scientific™
75008185 |
White |
Each for $228.04
|
|
|||||
75-008-186
|
Thermo Scientific™
75008186 |
Red |
Each for $228.04
|
|
|||||
75-008-188
|
Thermo Scientific™
75008188 |
Gray |
Each for $229.85
|
|
|||||
Description
- Accommodate tube sizes from 1.5 to 100mL, including urine tubes
Order Info
Adapters are sold individually; order correct quantity to equip rotor.
Specifications
4 x 180 Swing bucket rotor |