missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Microplate Carrier for H-6000A Buckets
Carriers for H-6000A, HBB-6, HLR-6 swinging bucket rotors
Supplier: Thermo Scientific™ 11267
Description
- Holds 6 x 3 standard or 6 x 1 deepwell microplates
- Maximum tube dimensions: 60 x 127mm
- Maximum Speed: 5000rpm
- Maximum RCF: 5186xG
Specifications
Microplate Carrier | |
H-6000A bucket | |
6 x 3 Standard or 6 x 1 Deepwell Microplates |