Thermo Scientific IEC Micro-MB Microcentrifuge, Microhematocrit Rotor
For 24 capillary tubes.
Manufacturer:  Thermo Scientific 002750F
Catalog No. 05-040-15
Description
Description
For use with: Micro-MB microcentrifuge
Capacity: 24 tubes, 1.75 dia. × 75mm long
Maximum speed/force: 11,500rpm/13,460×G
Specifications
Specifications
24 Capillary Tubes
11,500rpm
1.75mm
13,460 x g
75mm
Micro-MB Microcentrifuges
Documents