Thermo Scientific™ iD7 ATR Accessory for the Nicolet™ iS5
Get more from your analysis to meet your material identification and verification needs
Manufacturer:  Thermo Scientific™ 869172900
Catalog No. 14-387-800
Description
Description
The iD7 ATR Accessory for the Nicolet iS5 FT-IR Spectrometer provides the robustness and flexibility to meet a variety of material identification and verification needs. The optics of the iD7 ATR accessory maximize energy throughput, providing high-quality spectral data in seconds. Robust, simple operation is assured by the dependable crystal mounting system, and the Thermo Scientific™ Smart™ chip technology that integrates the iD7 ATR into your Nicolet iS5 spectrometer.
  • Monolithic diamond crystal provides superior robustness
  • Full spectral range provides more information about your sample
  • Interchangeable crystals offer sampling versatility
  • Automatic accessory recognition enables high productivity
Order Info
Requires an iD7 Crystal Plate for operation.
Available crystal types: diamond, coated (Mfr. No. 869-168800), diamond, uncoated (Mfr. No. 869-168900), ZnSe (Mfr. No. 869-169000), germanium (Mfr. No. 869-169100)
Specifications
Specifications
iD7 ATR Accessory
Documents