missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Thermo Scientific™ Open-top Tank Liner System, 200L
Designed for use with commercially available overhead mixers
Supplier: Thermo Scientific™ SH3064203
Description
Tank Liner Key Features:
- Fits into industry standard cylindrical tanks and in Thermo Scientific HyPerforma Conical Bottom Drums
- Gusset design allows for a close fit to the tank
- Chamber constructed of Thermo Scientific CX3-9 film, which provides the same contact surface as other Thermo Scientific HyPerforma BPCs
- Supplied gamma irradiated
- Customization options are available by contacting one of our Thermo Scientific Representatives today
Applications:
Hydration of powdered media and buffers; Pooling of nonsterile solutions and fluids
Order Info
Supplied double polybagged and irradiated with Certificate of Analysis.
Specifications
200 L | |
23 in. | |
1 | |
Gamma Irradiated | |
None | |
Top dispense | |
Non-sterile |
Plastic | |
Tank Liner | |
HyPerforma Conical Bottom Drum | |
182.9 cm (72 in) | |
Thermo Scientific | |
C-Flex |
For Research Use Only. Not for use in diagnostic procedures.