Thermo Scientific™ 384-Well Microtiter Microplates
Excellent for a number of applications.
$2117.59 - $2117.59
Specifications
Format Standard
Array 16 x 24
Product Type Microplate
Material Polystyrene
No. of Wells 384
View More Specs
Products 1
Catalog Number Mfr. No. Color Sterility Lid Price Quantity  
Catalog Number Mfr. No. Color Sterility Lid Price Quantity  
21-377-275
View Documents Promotion Details
Thermo Scientific™
95040000
Clear Non-sterile Without Lid
Case of 50 Each for $2,117.59
Description
Description
Excellent for a number of applications.
These microplates share the same footprint as 96-well versions.
  • Square wells
  • Design allows for miniaturization of assays and other applications
  • Use for high-volume DNA library manipulation and solid-phase binding
Use these microplates for the following application:
  • Clear plates—colorimetric applications
  • White plates—luminometric/fluorometric applications
  • Black plates—fluorometric applications
Specifications
Specifications
Standard
Microplate
384
Round Bottom (U-well)
50/Cs.
16 x 24
Polystyrene
Non-treated
For high throughput screening, PCR, mutagenicity/mutational spectra assays
SDS
missing translation for 'documents'
missing translation for 'documents'