Texwipe™ BetaMop™ II
Low contamination properties make this the perfect mop for floor cleaning in critical environments
$208.37 - $670.12
Catalog Number | Mfr. No. | Product Type | For Use With (Equipment) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | For Use With (Equipment) | Price | Quantity | ||||
19099196
|
Texwipe™
TX7106 |
BetaMop II Mop | Wring-bucket Systems |
Each for $208.37
|
|
||||
19099197
|
Texwipe™
TX716R |
Refill Mop Head | BetaMop II |
Case of 6 Each for $670.12
|
|
||||
Description
Durable, absorbent, corrosion-resistant, and low-contaminating; Autoclavable.
Durability, corrosion resistance, and low contamination properties make this the perfect mop for floor cleaning in critical environments. High-capacity, seamless, tubular-knit polyester mop head provides excellent absorbency. BetaMop II may be used with all wring-bucket systems. Both the mop head and clamp are autoclavable for use in sterile environments.
Specifications
Autoclavable | |
Corrosion | |
Polyester |