Learn More
Tetramethylgermanium, 98%, Thermo Scientific Chemicals
$436.25 - $1728.38
Chemical Identifiers
CAS | 865-52-1 |
---|---|
Molecular Formula | C4H12Ge |
Molecular Weight (g/mol) | 132.77 |
MDL Number | MFCD00014843 |
InChI Key | ZRLCXMPFXYVHGS-UHFFFAOYSA-N |
Synonym | tetramethylgermanium, germanium, tetramethyl, germane, tetramethyl, tetramethyl germane, tetramethyl germanium, acmc-1bk8h, germanium tetramethyl, ch3 4ge, 4-04-00-04282 beilstein handbook reference |
PubChem CID | 70079 |
IUPAC Name | tetramethylgermane |
SMILES | C[Ge](C)(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1693106
|
Thermo Scientific Chemicals
A1693106 |
5 g |
Each for $436.25
|
|
|||||
AAA1693114
|
Thermo Scientific Chemicals
A1693114 |
25 g |
Each for $1,728.38
|
|
|||||
Description
Employed as a Chemical Vapor Deposition (CVD) precursor to germanium oxide thin films.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEmployed as a Chemical Vapor Deposition (CVD) precursor to germanium oxide thin films.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
Chemical Identifiers
865-52-1 | |
132.77 | |
ZRLCXMPFXYVHGS-UHFFFAOYSA-N | |
70079 | |
C[Ge](C)(C)C |
C4H12Ge | |
MFCD00014843 | |
tetramethylgermanium, germanium, tetramethyl, germane, tetramethyl, tetramethyl germane, tetramethyl germanium, acmc-1bk8h, germanium tetramethyl, ch3 4ge, 4-04-00-04282 beilstein handbook reference | |
tetramethylgermane |
Specifications
865-52-1 | |
0.975 | |
−37°C (−35°F) | |
1.389 | |
5 g | |
tetramethylgermanium, germanium, tetramethyl, germane, tetramethyl, tetramethyl germane, tetramethyl germanium, acmc-1bk8h, germanium tetramethyl, ch3 4ge, 4-04-00-04282 beilstein handbook reference | |
ZRLCXMPFXYVHGS-UHFFFAOYSA-N | |
tetramethylgermane | |
70079 | |
98% |
-88°C | |
42°C to 44°C | |
C4H12Ge | |
MFCD00014843 | |
UN1993 | |
Not miscible or difficult to mix in water. | |
C[Ge](C)(C)C | |
132.77 | |
132.73 | |
Tetramethylgermanium |
Safety and Handling
GHS H Statement
H225-H302-H312-H332
Highly flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P363-P370+P378q-P501c
H224-H302+H312+H332-H319
DOTInformation : Hazard Class: 3; Packaging Group: I
EINECSNumber : 212-745-0
RTECSNumber : LY5345000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only