missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrahydrofuran, Anhydrous, 99.9% min., MilliporeSigma™

$110.92 - $211.92

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MTX02777
View Documents Promotion Details
Technidata
TX02777
100 mL
Each for $110.92
 
MTX02776
View Documents Promotion Details
Technidata
TX02776
1 L
Each for $211.92
 
Description

Description

Specifications

Chemical Identifiers

109-99-9
72.11
WYURNTSHIVDZCO-UHFFFAOYSA-N
8028
oxolane
C4H8O
MFCD00005356
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
CHEBI:26911
C1CCOC1

Specifications

-108.3°C
0.89g/cm3 at 20°C
100 mL
320.85°C (609.5°F)
Liquid
C4H8O
MFCD00005356
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
WYURNTSHIVDZCO-UHFFFAOYSA-N
oxolane
8028
72.11
109-99-9
66.1°C
15.15°C (4.7°F), Closed cup
Ether-like
99.9%, min.
CH2(CH2)3O
UN 2056
Soluble in water
C1CCOC1
72.11
CHEBI:26911
Tetrahydrofuran
Safety and Handling

Safety and Handling

DOTInformation : DOT Class: 3, Packaging Group: II

Recommended Storage : Room Temperature

SDS
missing translation for 'documents'

missing translation for 'documents'