missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Troemner™ Talboys™ Basic Mini Hotplate-Stirrer

Ergonomic compact unit that heat and stir up to 1000mL of liquid.

Manufacturer:  Troemner™ 984TA5AHSUS

Catalog No. S14174



Description

Description

  • New Ergonomic Design
  • Ideal for educational labs
  • Boils 300mL of water in 16 minutes
  • Aluminum top plate 12.7 x 12.7cm (5 x 5 in.) for even heating surface
  • Supplied with 183cm (72 in.), 3-wire cord and plug and 3.8cm (1.5 in.) PTFE coated stir bar
  • Built in support rod holder with locking knob accepts optional Support Rod and Clamp Kit

Applications:

General heating, boiling, chemical reactions, evaporations, digestions, disolving reagents, buffers, and dehydrated media, concentrations, oil bath, titrations, dissolutions, general mixing

Certifications

TUV

Warranty

5-Year

Specifications

Specifications

33.81 oz.
100 to 1200rpm
375°C
6A
585w
12.7 x 12.7cm
120V 50/60Hz
Top Plate: Aluminum
Up to 400°C
1000mL
707°F
120V
50/60Hz
5 x 5 in.
33.81 oz., 1000mL
Up to 400°C
1000mL
5 Years
Documents