Kent Scientific SurgiSuite and Lighting and Mouse Intubation Kit Package
Simple to order, economical package to perform surgical applications, which includes SurgiSuite multi-functional surgical platform kit, lighting kit and mouse intubation kit.
Manufacturer:  Kent Scientific SURGIETI
Catalog No. 13005183
Includes:
SurgiSuite Multi-functional Surgical Platform:
One chemical resistant base, one stainless steel surgical surface, one rechargeable far infrared warming pad, two magnetic anesthesia mask/intubation tubing stabilizers (one small and one large), fou
Description
Description
SurgiSuite Multi-functional Surgical Platform:
  • Ensures animal warming and homeothermic control
  • Integrated warming by rechargeable, battery-operated far infrared warming pad
  • Multi-positional stabilizers
  • Platforms are surrounded by ferro-magnetic stainless steel plate
  • Incorporating window allows warming of animal during surgical procedure
Lighting Kit:
  • Provides ambient lighting to surgical set up
Mouse Intubation Kit:
  • Integrated anesthesia mask delivers isoflurane during intubation
  • Independent neck positioner aligns airway structures
  • Fiber optic light illuminates airway for enhanced visualization
Specifications
Specifications
Surgery, homeothermic warming, surgical procedures, tissue retraction
SurgiSuite and Lighting and Mouse Intubation Kit Package
Documents