Thermo Scientific™ SUN-SRi™ Magnetic Screw-Thread Closures
$124.52 - $189.94
Specifications
Type Screw Cap
Thread Size 18mm
View More Specs
Products 3
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
22-030206 View Documents Thermo Scientific™
405035
Pack of 100 Each for $174.33
22-030204 View Documents Thermo Scientific™
405029
Pack of 100 Each for $124.52
22-030205 View Documents Thermo Scientific™
405031
Pack of 100 Each for $189.94
Description
Description
  • 18mm closures fit both the 10 and 20mL screw-top headspace vials
  • Can be used with Gerstel™, CTC/Leap™ Technologies, and other magnetic transport autosamplers
Specifications
Specifications
Screw Cap
18mm