missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

HemoCue America StatSpin™ MP Multipurpose Centrifuge

For microhematocrits, urine, sedimentation and blood sample preparation

Manufacturer:  HemoCue America SSMP

Catalog No. 22-314934



Includes: Centrifuge, 12-place hematocrit rotor and reader, two-place tube rotor for urine and blood tubes, vial of hematocrit tubes (22-269-245) and Hematocrit Sealant Pad (22-269-247)

Description

Description

  •  only supports 40mm tube lengths

Order Info

Shipping weight: 9 lb. (4.1kg)

Specifications

Specifications

12,999 x G
No
12,000 to 12,999 x G
Yes
29,999rpm
2-place Tube Rotor
20,000 to 29,999rpm
Documents
Accessories