missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Starch Solution (Stabilized), Fisher Chemical

For iodometry

$118.85 - $223.00

Specifications

Chemical Name or Material Starch Solution
Name Note Stabilized
CAS 9005-25-8,64-19-7,7732-18-5
Molecular Formula (C6H10O5)n
MDL Number MFCD00082026
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
SS408-1 View Documents Thermo Scientific
SS4081
1L Poly Bottle Each for $118.85
Case of 6 Each for $713.10
SS408-4 View Documents Thermo Scientific
SS4084
4L Poly Bottle Each for $223.00
Case of 4 Each for $892.00
Specifications

Specifications

Starch Solution
9005-25-8,64-19-7,7732-18-5
MFCD00082026
Stabilized
(C6H10O5)n