missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Standard, Water, 1%, Apura™, EMD Millipore

Manufacturer:  Millipore Sigma 1.88052.0010

Catalog No. M1880520010


 

Specifications

Specifications

Standard, Water
Apura™ Karl Fischer Water Standard, Mixture. Contains 1-Methoxy-2-propanol.
1.00%
8 x 10mL
Glass Ampoule
Contains 1-Methoxy-2-propanol

Chemical Identifiers

1.00%
Standard, Water, Apura™ Karl Fischer Water Standard, Mixture. Contains 1-Methoxy-2-propanol.
Safety and Handling

Safety and Handling

Recommended Storage : Store without restrictions

SDS Documents