missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mettler Toledo™ Standard Line Weight Set: 1mg-200g E2 AL

Standard Line Weights. Specially used stainless steel, including the mg range, ensures anti corrosive surfaces with low magnetization and susceptibility values. Solid, one piece construction and mirror-polished surface grades give stable values.

Manufacturer:  Mettler Toledo™ 30014234

Catalog No. 01-914-437


 

Description

Description

  • Box: Plastic box (included)
  • Calibration Certificate: No
  • Content (set up): 1 mg - 200 g (23 pieces)
  • Density p: 7950 (± 140) kg/m&superscript_3;
  • Design: One-piece (monobloc)
  • Material: Stainless Steel
  • Finest and pure stainless steel - Specially used stainless steel, including the mg range, ensures anti corrosive surfaces with low magnetization and susceptibility values.
  • Solid, one-piece construction - The one piece construction and mirror-polished surface grades give stable values.
  • Weight calibration performed by robots - Substantial investments in cutting-edge robot technology, combined with decades of experience have opened up a new field in weight calibration.
Specifications

Specifications

Standard Line Weight Set
Stainless Steel
Balance calibration and testing
Class E2
1mg-200g
Documents