missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mettler Toledo™ Standard Line Weight: 10g E2 PL, Calib. Certificate

Standard Line Weights. Specially used stainless steel, including the mg range, ensures anti corrosive surfaces with low magnetization and susceptibility values. Solid, one piece construction and mirror-polished surface grades give stable values.

Manufacturer:  Mettler Toledo™ 30375804

Catalog No. 01-914-425



Description

Description

  • Box: Plastic box (included)
  • Calibration Certificate: Yes
  • Density p: 7950 (± 140) kg/m&superscript_3;
  • Material: Stainless Steel
  • OIML Class: E2
  • Susceptibility X: E2 ≤ 0.0049
  • Finest and pure stainless steel - Specially used stainless steel, including the ones in the mg range, ensures anti corrosive surfaces with low magnetization and susceptibility values
  • Solid, one-piece construction - The one piece construction and mirror-polished surface grade results in stable values
  • Weight calibration performed by robots - Substantial investments in cutting-edge robot technology combined with decades of experience have opened up a new field in weight calibration
Specifications

Specifications

Standard Line Weight
Stainless Steel
Balance calibration and testing
Class E2
10g
Documents