Thermo Scientific™ Standard Fluid Transfer Assembly Adapter Set
Simplify process connectivity and enable greater efficiency for most upstream and downstream process operations.
$31.06 - $60.15
Specifications
Storage Requirements Approved for storage at ambient conditions (15°C to 30°C).
View More Specs
Products 8
Catalog Number Mfr. No. Tubing Style Price Quantity  
Catalog Number Mfr. No. Tubing Style Price Quantity  
SH3108401 View Documents Thermo Scientific™
SH3108401
3.2mm (1/8 in) x 1.6mm (1/16 in) wall to 6.35mm (1/4 in) ID x 1.6mm (1/16 in) wall, C-Flex 374, 2m (20 in) Each for $31.06
SH3108501 View Documents Thermo Scientific™
SH3108501
3.2mm (1/8 in) x 1.6mm (1/16 in) wall to 6.35mm (1/4 in) ID x 2.4mm (3/32 in) wall, C-Flex 374, 2m (20 in) Each for $32.06
SH3108601 View Documents Thermo Scientific™
SH3108601
3.2mm (1/8 in) x 1.6mm (1/16 in) wall to 9.52mm (3/8 in) ID x 3.2mm (1/8 in) wall, C-Flex 374, 2m (20 in) Each for $33.26
SH3108701 View Documents Thermo Scientific™
SH3108701
6.35mm (1/4 in) x 1.6mm (1/16 in) wall to 9.52mm (3/8 in) ID x 3.2mm (1/8 in) wall, C-Flex 374, 2m (20 in) Each for $33.69
SH3108801 View Documents Thermo Scientific™
SH3108801
6.35mm (1/4 in) x 1.6mm (1/16 in) wall to 12.7mm (1/2 in) ID x 3.2mm (1/8 in) wall, C-Flex 374, 2m (20 in) Each for $60.15
SH3108901 View Documents Thermo Scientific™
SH3108901
6.35mm (1/4 in) x 2.4mm (3/32 in) wall to 9.52mm (3/8 in) ID x 3.2mm (1/8 in) wall, C-Flex 374, 2m (20 in) Each for $42.11
SH3109001 View Documents Thermo Scientific™
SH3109001
6.35mm (1/4 in) x 2.4mm (3/32 in) wall to 12.7mm (1/2 in) ID x 3.2mm (1/8 in) wall, C-Flex 374, 2m (20 in) Each for $36.15
SH3109101 View Documents Thermo Scientific™
SH3109101
9.52mm (3/8 in) x 3.2mm (1/8 in) wall to 12.7mm (1/2 in) ID x 3.2mm (1/8 in) wall, C-Flex 374, 2m (20 in) Each for $44.57
Description
Description
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process. Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
  •  One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
  • Multiple manufacturing sites ensure security in product supply
  • Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
  • Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Specifications
Specifications
Approved for storage at ambient conditions (15°C to 30°C).