missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Stainless Steel Foil, 0.25mm (0.01 in.) thick, Thermo Scientific Chemicals
$89.14 - $89.14
Specifications
Chemical Name or Material | Stainless Steel |
---|---|
Color | Silver |
Appearance | Silver -colored foil |
Molecular Formula | Fe:Cr:Ni; 70:19:11 wt% |
MDL Number | MFCD00148499 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA41584GH
|
Thermo Scientific Chemicals
041584GH |
100 x 100 mm |
Each for $89.14
|
|
|||||
Description
It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Stainless Steel | |
Silver -colored foil | |
MFCD00148499 | |
Odorless |
Silver | |
Fe:Cr:Ni; 70:19:11 wt% | |
It is insoluble in water. | |
Foil |
RUO – Research Use Only