missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Spectrum™ Spectra/Por™ 4 RC Dialysis Membrane Tubing 12,000 to 14,000 Dalton MWCO
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting.
$570.85 - $669.27
Specifications
Product Type | Spectra/Por 4 Dialysis Tubing |
---|---|
MWCO | 12 to 14 kDa |
Material | Regenerated Cellulose |
Length (English) | 100 ft. |
Formulation | Packaged with glycerin |
Catalog Number | Mfr. No. | Diameter (English) | Diameter (Metric) | Length (Metric) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (English) | Diameter (Metric) | Length (Metric) | Price | Quantity | |||
08-667E
|
Spectrum™
S432706 |
1.77 in. | 45 mm | 30 m |
N/A
|
|
|||
08-667B
|
Spectrum™
S432700 |
0.62 in. | 16 mm | 30.5 m |
N/A
|
|
|||
Description
Supplied as flat-width tubing or flat sheets that can be folded to fit various funnel-type dialysis apparatus.
Pure regenerated natural cellulose membranes with 1000 to 50,000 molecular weight cutoffs.
- Transparent and flexible
- Low molecular weight compounds in aqueous solutions diffuse through membrane
- Higher molecular weights, such as proteins and bacteria, are retained by membrane
- Glycerol preserves membrane and must be removed before use
- Flat sheets and discs available for use with dialysis apparatus
Do not tie knots in tubing; use closures
Specifications
Spectra/Por 4 Dialysis Tubing | |
Regenerated Cellulose | |
Packaged with glycerin | |
4°C to 121°C | |
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting |
12 to 14 kDa | |
100 ft. | |
39.2°F to 249.8°F | |
Each |