Sonoco™ ThermoSafe Insulated EPS Foam Only Containers and Coolers (Medium)
Large selection of cost-effective, reusable EPS foam only containers available in small, medium, and large sizes. EPS foam only shippers for the safe storage and delivery of most refrigerated or frozen products.
Supplier:  Sonoco™ TS452
 View more versions of this product
Catalog No. 03-528-18
Description
Description
  • Sizes that range 11 inches-17 inches in length
  • Also available in ThermoSafe Insulated EPS Foam Only Containers and Coolers- Small (6  inches-10.5 inches in length)
  • Also available in ThermoSafe Insulated EPS Foam Only Containers and Coolers- Large (19inches-24 inches in length)
  • Excellent dry ice sublimation… average dry ice usage in 1-1/2 inch thick wall units is 5 pounds over 24 hours and 3-4 pounds thereafter.
  • Standard 1-1/2 inch wall shippers are ideal for refrigerated shipments for 24-48 hours.
Specifications
Specifications
3 per case
16 X 16 X 15.5 in.
16 in.
12 X 12 X 11.5 in.
Insulated Shipper Container
16L x 16W x 15.5 in. H
12L x 12W x 11.5 in. H
Foam