missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium thiosulfate, 0.1 N standard solution, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 124270025

 View more versions of this product

Catalog No. AC124270025



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent, Volumetric standard solution
Specifications

Specifications

Sodium thiosulfate
7772-98-7,7732-18-5
2.5 L
Na2S2O3
15, 8821
Solubility in water: soluble in water. Other solubilities: practically insoluble in alcohol
[Na+].[Na+].[O-]S([S-])(=O)=O
158.10
158.11
Plastic Bottle
Colorless
Liquid
Na2O3S2
MFCD00003499
sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt
AKHNMLFCWUSKQB-UHFFFAOYSA-L
disodium sulfanidesulfonate
24477
0.0950 to 0.1050N (20°C)
SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only