missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Sulfate Anhydrous, min.99%, 12-60 Mesh, Granular, Ultra Resi-Analyzed, ACS Reagent, J.T. Baker™
Supplier: Avantor 337501
Specifications
Sodium Sulfate | |
7757-82-6 | |
Granules | |
Meets ACS reagent chemical requirements | |
MFCD00003504 | |
PMZURENOXWZQFD-UHFFFAOYSA-L | |
disodium;sulfate | |
24436 | |
142.04 | |
max. 0.5% | |
1L = 2.68kg |
Anhydrous | |
5.2 to 9.2 | |
500 g | |
Na2O4S | |
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 | |
[Na+].[Na+].[O-]S([O-])(=O)=O | |
142.04 | |
CHEBI:32149 | |
ACS Reagent | |
Wide Mouth Amber Glass Bottle |
Chemical Identifiers
7757-82-6 | |
142.04 | |
PMZURENOXWZQFD-UHFFFAOYSA-L | |
24436 | |
[Na+].[Na+].[O-]S([O-])(=O)=O |
Na2O4S | |
MFCD00003504 | |
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 | |
CHEBI:32149 |