missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical

For atomic absorption. (1mL = 1mg Na) Solute: Sodium Chloride Solvent: Distilled Water

$89.82 - $95.59

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00085307
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium;chloride
SMILES [Na+].[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
SS139-500 SDS View Documents Thermo Scientific
SS139500
500mL Poly Bottle Each for $89.82
Case of 6 Each for $538.92
SS139-100 SDS View Documents Thermo Scientific
SS139100
100mL Poly Bottle Each for $95.59
Case of 6 Each for $573.54
Specifications

Chemical Identifiers

7647-14-5
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
CHEBI:26710
[Na+].[Cl-]
ClNa
MFCD00085307
5234
sodium;chloride

Specifications

7647-14-5,7732-18-5
MFCD00085307
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium;chloride
5234
Liquid
Poly Bottle
14mmHg
0°C
Colorless
Sodium, Reference Standard Solution, 1000ppm ±1%
ClNa
990 to 1010ppm
[Na+].[Cl-]
58.44
CHEBI:26710
Certified
LOT NUMBER USED SODIUM REF. STD.
1.0g/cm³
100°C
500mL
Safety and Handling

Safety and Handling