missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium hydroxide, pure, 1N Standard Solution, Thermo Scientific Chemicals
$63.98 - $183.22
Specifications
Chemical Name or Material | Sodium hydroxide |
---|---|
Color | Colorless |
CAS | 1310-73-2,7732-18-5 |
Molecular Formula | HNaO |
Linear Formula | NaOH |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC124260010
|
Thermo Scientific Chemicals
124260010 |
1 L | Plastic Bottle |
Each for $63.98
|
|
||||
AC124260050
|
Thermo Scientific Chemicals
124260050 |
5 L | Plastic drum |
Each for $183.22
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Volumetric standard solutionSpecifications
Sodium hydroxide | |
1310-73-2,7732-18-5 | |
NaOH | |
01,1083; 05,616; 07,336; 08,460 | |
15, 8761 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
40 |
Colorless | |
HNaO | |
0.998 to 1.002N (20°C) | |
1.0400g/mL | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
Liquid |
RUO – Research Use Only