missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Hydroxide, 1.00 N, Ricca Chemical
Standardized Bases
$64.06 - $469.97
Chemical Identifiers
CAS | 1310-73-2 |
---|---|
Molecular Formula | HNaO |
Molecular Weight (g/mol) | 39.997 |
InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
PubChem CID | 14798 |
ChEBI | CHEBI:32145 |
IUPAC Name | sodium;hydroxide |
SMILES | [OH-].[Na+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
7450-4
|
Ricca Chemical Company
74504 |
120 mL | Natural Poly Bottle |
Each for $65.34
|
|
||||
7450-16
|
Ricca Chemical Company
745016 |
500 mL | Natural Poly Bottle |
Each for $64.06
|
|
||||
R74500001F
|
Ricca Chemical Company
R74500001F |
1 L | Cubitainer |
Each for $84.78
|
|
||||
7450-32
|
Ricca Chemical Company
745032 |
1 L | Natural Poly Bottle |
Each for $75.66
|
|
||||
74501CT
|
Ricca Chemical Company
74501CT |
4 L | Cubitainer |
Each for $250.84
|
|
||||
7450-1
|
Ricca Chemical Company
74501 |
4 L | Natural Poly Bottle |
Each for $171.75
|
|
||||
7450-25
|
Ricca Chemical Company
745025 |
10 L | Cubitainer |
Each for $348.25
|
|
||||
7450-5
|
Ricca Chemical Company
74505 |
20 L | Cubitainer |
Each for $469.97
|
|
||||
Chemical Identifiers
1310-73-2 | |
39.997 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
CHEBI:32145 | |
[OH-].[Na+] |
HNaO | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
14798 | |
sodium;hydroxide |
Specifications
1310-73-2,7732-18-5 | |
94.0,7.54 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
ACS/USP Volumetric | |
Natural Poly Bottle |
91.0,7.25 | |
HNaO | |
120 mL | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
1.00N | |
Sodium Hydroxide |
Safety and Handling
ShelfLife : 24 months
Recommended Storage : Ambient Corrosive