missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Hydroxide, 1.00 N, Ricca Chemical
Standardized Bases
Supplier: Ricca Chemical Company 745025
Specifications
Sodium Hydroxide | |
91.0,7.25 | |
HNaO | |
10 L | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
1.00N |
1310-73-2,7732-18-5 | |
94.0,7.54 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
ACS/USP Volumetric | |
Cubitainer |
Chemical Identifiers
1310-73-2 | |
39.997 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
CHEBI:32145 | |
[OH-].[Na+] |
HNaO | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
14798 | |
sodium;hydroxide |