missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Carbonate, Anhydrous, ACS Grade, ≥ 99.5%, LabChem™
Supplier: LabChem LC229651
Specifications
Sodium Carbonate | |
497-19-8 | |
100 | |
500 g | |
Na2CO3 | |
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
disodium carbonate | |
10340 | |
105.99 | |
ACS | |
Poly Bottle | |
Fume; Carbon monoxide; Carbon dioxide |
Anhydrous | |
White | |
Powder | |
CNa2O3 | |
MFCD00003494 | |
Soluble in water | |
[Na+].[Na+].[O-]C([O-])=O | |
105.99 | |
CHEBI:29377 | |
≥99.5% | |
Passes Test | |
2.53g/cm3 | |
2.53g/cm3 |
Chemical Identifiers
497-19-8 | |
105.99 | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
10340 | |
disodium carbonate |
CNa2O3 | |
MFCD00003494 | |
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
CHEBI:29377 | |
[Na+].[Na+].[O-]C([O-])=O |
Safety and Handling
GHS H Statement
Causes skin irritation and serious eye irritation.
GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Warning
EINECSNumber : 207-838-8
Recommended Storage : Room Temperature