missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silicon oil 550, EMD-Millipore

Manufacturer:  Millipore Sigma 1097620100

Catalog No. M1097620100



Specifications

Specifications

Silicon oil 550
Glass
Density: 1.07g/cm3 (20°)
100mL
>400°C DIN 51794
208mm2/sec., kinematic (20°C)
Solvent: Chloroform
Documents