Learn More
Silicon carbide, alpha-phase, 99.8% (metals basis), Thermo Scientific Chemicals
Surface Area 9-11 m2/g
$123.68 - $1141.97
Chemical Identifiers
CAS | 409-21-2 |
---|---|
Molecular Formula | CSi |
Molecular Weight (g/mol) | 40.10 |
MDL Number | MFCD00049531 |
InChI Key | HBMJWWWQQXIZIP-UHFFFAOYSA-N |
Synonym | silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide |
PubChem CID | 9863 |
ChEBI | CHEBI:29390 |
SMILES | [C-]#[Si+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA4015522
|
Thermo Scientific Chemicals
04015522 |
100 g |
Each for $123.68
|
|
|||||
AA4015536
|
Thermo Scientific Chemicals
04015536 |
500 g |
Each for $328.90
|
|
|||||
AA40155A3
|
Thermo Scientific Chemicals
040155A3 |
2 kg |
Each for $1,141.97
|
|
|||||
Description
Used in abrasives, in polishing and grinding. It is widely applied in applications calling for high endurance, such as automobile brakes, car clutches and ceramic plates in bulletproof vests. Electronic applications of silicon carbide are as light emitting diodes and sensors. It is utilized for the production of fertile, transgenic maize plants. Silicon carbide whiskers and zirconium diboride ceramic. It compositely plays an important role in the crack-healing behavior.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
409-21-2 | |
40.10 | |
HBMJWWWQQXIZIP-UHFFFAOYSA-N | |
9863 | |
[C-]#[Si+] |
CSi | |
MFCD00049531 | |
silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide | |
CHEBI:29390 |
Specifications
2700°C | |
409-21-2 | |
100 g | |
CSi | |
silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide | |
HBMJWWWQQXIZIP-UHFFFAOYSA-N | |
methanidylidynesilylium | |
9863 | |
40.1 | |
Silicon carbide, alpha-phase |
Black to Gray | |
Powder | |
99.8% (metals basis) | |
MFCD00049531 | |
Soluble in molten sodium hydroxide,potassium hydroxide and in molten iron. | |
[C-]#[Si+] | |
40.10 | |
CHEBI:29390 | |
Odorless |
Safety and Handling
H351-H315-H319-H335: Suspected of causing cancer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
EINECSNumber : 206-991-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only